Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Boardwalk® Light Duty Scour Pad
Supplier: Boardwalk
Boardwalk®
BWK198
21899-525CS 34.55 USD InStock
21899-525
Boardwalk® Light Duty Scour Pad
Wipes Cleaning Kits and Pads
Ideal for cleaning glass, stainless steel, porcelain, chrome, copper and ceramic surfaces.
Won't scratch or dull most surfaces.
Ordering information: Includes 20 pads per carton.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty