Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Feather® Sterile Surgical Blade, GF Health Products (Graham-Field)
Supplier: GF Health Products (Graham-Field)
2976#11
21899-530BX 89.62 USD InStock
21899-530
Feather® Sterile Surgical Blade, GF Health Products (Graham-Field)
Scalpels and Scalpel Assemblies Scalpel Blades
Superior cutting quality, strength and corrosion resistance characterize these stainless steel, disposable blades.
  • Individually wrapped and sealed in aluminum foil to maintain sterility, insure long shelf-life
  • Blades available in several sizes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty