Would you like to log in again?
Boardwalk® Stripping Floor Pads
Supplier: Boardwalk
Boardwalk®
BWK4017BLA
21899-532CS 82.73 USD InStock
21899-532
Boardwalk® Stripping Floor Pads
Wipes Cleaning Kits and Pads
For long-lasting stripping of finish, dirt and buildup.
  • For stripping finished floors
  • 1" in height
Use with rotary machines (speed of 175 to 600 rpm). Color-coded for easy job identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty