Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Boardwalk® Buffing Floor Pads

Supplier: Boardwalk
Boardwalk®
BWK4017RED
21899-534CS 82.71 USD InStock
21899-534
Boardwalk® Buffing Floor Pads
Wipes Cleaning Kits and Pads
Cleans and removes scuff marks while polishing floors to a high gloss.

  • Cleans when wet, buffs when dry
  • For use with Rotary Machines (speed of 175 to 600 rpm)
  • Height: 1"

Color-coded for easy job identification. For finished floors.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty