Would you like to log in again?
Process NPD® One-Step Germicidal Detergents
Supplier: Steris
Process NPD®
639008 639001 639002
77531-582EA 3138.13 USD InStock
77531-582 21899-728 77531-580
Process NPD® One-Step Germicidal Detergents
Cleaning Agents Detergents and Surface Cleansers
STERIS' Process NPD® One-Step germicidal detergent is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' Process NPD® One-Step Germicidal Detergent is the non-sterile version of the Process NPD® family of one-step, quaternary ammonium cleaner/disinfectants made specifically for use on hard non-porous surfaces. The Process NPD® products are all made with the same formulation that contains no perfumes or dyes that might interfere with critical research studies or manufacturing processes. At a use-dilution rate of 1:256, the Process NPD® products kill a broad spectrum of common bacteria without harming the surface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty