Would you like to log in again?
Glass-Klenz® Glassware Detergentz, STERIS®
Supplier: Steris
Glass-Klenz®
111408
21899-881CS 439.92 USD InStock
21899-881
Glass-Klenz® Glassware Detergentz, STERIS®
Cleaning Agents Detergents and Surface Cleansers
Glass-Klenz® Glassware detergent is a low-foaming, chelant-based, liquid detergent for cleaning laboratory glasswares.
  • Effective removal of laboratory residues in both soft and hard water
  • Effectively holds soils in suspension allowing to be completely removed without redeposition
  • Reduces utility consumption and cleaning time due
  • Mild pH for enhanced safety and substrate compatibility
It is a free rinsing detergent specially formulated for use in automated glassware washing applications for the removal of particulate.
Certifications: Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty