Would you like to log in again?
KIMTECH PURE® W4 Critical Task Wipers, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
33390 33330 33390 33330
21908-205CS 333.15 USD InStock
21908-205 CA21908-206 83009-302 CA83009-302
KIMTECH PURE® W4 Critical Task Wipers, Kimberly-Clark Professional®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Low-linting wipers for ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
  • 100% polypropylene, non-woven fabric is low in particulate matter and extractable levels
  • Soft, absorbent, durable
  • Contain no binders or reinforcing fibers
  • Eliminiate surface fibers, trap particles, will not scratch surfaces
  • Color: white
Each wiper can absorb over four times its weight in liquids. Acid-, base-, and solvent-resistant; will not break down or disintegrate when exposed to chemicals.
For ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom assembly operations, pharmaceutical operations, and other clean environments.
Certifications: Conform to EIA 541 standard. Meet the USDA requirements for food contact.
Ordering information: Packaged in antistatic, self-dispensing double poly bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty