Would you like to log in again?
Jameson Caliper, Integra® Miltex®
Supplier: Integra LifeSciences
18-658
21909-476EA 357.05 USD InStock
21909-476
Jameson Caliper, Integra® Miltex®
Calipers
Graduated from 0-80 mm in 1 mm increments
Stainless steel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty