Would you like to log in again?
Surgical Instrument Lubricants, Integra™ Miltex®
Supplier: Integra LifeSciences
3-705 3-700
95042-714EA 86.8 USD InStock
95042-714 21909-538
Surgical Instrument Lubricants, Integra™ Miltex®
Scissors
Designed to extend the working life of surgical instruments, this lubricant is pre-mixed, non-silicone based, and anti-corrosive
The water emulsible soaking agent is formulated with gentle, non-aggressive ingredients, and can be used daily to prevent spotting, staining, and rusting. Lubricant can be used on instrument box locks, screw locks, scissor blades, and any other metal on metal moving part. Available in an 8oz. spray pump bottle or 1gal. jug with dispensing pump.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty