Would you like to log in again?
Cheesecloth Wipes, Fiberweb
Supplier: Fiberweb
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
589929 589922 589923
21910-118CS 2625.6 USD InStock
21910-118 21910-130 21910-121
Cheesecloth Wipes, Fiberweb
Wipes Reusable Cleaning Cloths Cloths for Delicate Tasks
These cheesecloth wipes have an ideal absorbent surface for applying a uniform, thin film of gum arabic to an offset plate.
The wipes also prevent scratching or scuffing of the image area. They allow for quick roll up and increased water receptivity in the non-image areas, reducing scumming and halftone plugging and requiring less water, particularly at startup.
Economy-cut rolls 21910-118, -121 have 120 4-ply pieces per roll. Handi-fold bolts 21910-127, -124, and -130 are continuous Z-folded stacks with approximately 45.7 cm (18") between each fold.
Gum arabic, an acidic water-soluble gum from the acacia tree, adheres to the metal surface or plate and protects the non-image areas from oxidation, scratches, grease, and dirt. These cheesecloth wipes are excellent for buffing gummed plates to a dry, smooth, and streak-free finish with no linting or residue. They properly desensitize plates to ensure the best possible performance.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty