Would you like to log in again?
MicroFirst® LP Wipers, Berkshire
Supplier: Berkshire
MicroFirst®
MFLP.0909.12
21913-194CS 1349.42 USD InStock
21913-194
MicroFirst® LP Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These extremely clean and pure polyester/rayon non-woven wipers are made from soft, absorbent Dupont™ Sontara® MicroPure LP fabric.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of lyocell
  • Exceptionally pure and absorbent
  • Very low particle and fiber generation
  • Very durable with high wet strength
Because this fabric has no binders or chemical treatments, its cleanliness level rivals that of cleanroom-laundered, knitted synthetic wipers. Ideal for wiping delicate surfaces and packaging sensitive components. Double-bagged to ensure cleanliness.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty