Would you like to log in again?
MicroSeal® 1200 Wipers, Berkshire
Supplier: Berkshire
MicroSeal®
MS12000909.8 MS12001212.6 MS1200404B10
21913-198CS 1113.55 USD InStock
21913-198 21913-199 47732-718
MicroSeal® 1200 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
MicroSeal® 1200 wipers are designed for spill control, equipment cleaning, and general cleanup in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
  • Critically low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, Acetone, and other solvents
  • Weight and caliper provide an excellent hand for cleaning rough surfaces
Continuous-filament, double-knitted, 100% polyester fabric is highly absorbent and tear-resistant. Wipers have clean, pliable, sealed edges.
Ordering information: Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom. Wipers are stacked and double-bagged. Packs are individually labeled for full traceability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty