Would you like to log in again?
Durx® 770 Wipers, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Durx®
DR7701212.20 DR7700909.20 DR770040440 DR770.1818.10
10810-626CS 846.72 USD InStock
10810-626 21913-373 21913-351 21913-395
Durx® 770 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for absorbing spills of solvents and deionized water in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
  • Excellent combination of synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Reduced particle and fiber generation with ultra-low particle and fiber treatment
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
  • Smooth, absorbent, and durable
Also suitable for general surface and equipment cleaning. Nonwoven wipers are made from a blend of 45% polyester and 55% cellulose with superior strength and absorbency - wipers are able to absorb four times their own weight. Manufactured without binders or surfactants. Proprietary treatment enhances cleanliness by reducing particle generation. Low levels of ions and organic extractables.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty