Would you like to log in again?
Durx® 670 Wipers, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Durx®
DR6701212.20 DR6700707.45 DR6700909.20 DR670.082410 DR6700404.10 DR6701818.10
94000-404CS 462.05 USD InStock
94000-404 21913-410 21913-453 21913-431 21913-475 21913-412
Durx® 670 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for absorbing spills of solvents and deionized water in the cleanroom.
  • Excellent combination of synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
Also for general surface and equipment cleaning. Non-woven wipers are made from a polyester/cellulose blend of superior strength and absorbency, with low levels of ions and organic extractables. Made without binders or surfactants, resulting in a pure wiper.
Certifications: Certified for contact with food per EC Regulation 1935(2004) - FDA 21 CFR - Food contact compliant. Suitable for ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty