Would you like to log in again?
Twillx® 1622 Wipers, Berkshire
Supplier: Berkshire
Twillx®
T1622.0909.8 T1622.1212.4
21914-106CS 2185.15 USD InStock
21914-106 21914-128
Twillx® 1622 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durable 100% cotton twill wipers for cleaning and burnishing of rigid, grooved, slotted, or hard-to-reach surfaces.
  • Excellent strength and durability
  • Good absorbency
  • High heat resistance
  • Excellent resistance to the buildup of electrostatic discharge in 40 to 60% RH environments
  • Works well with IPA and other solvents
  • Autoclavable
Ideal for heavy-duty wiping and cleanup of diffusion furnaces. Natural cotton fibers provide excellent absorbency and thermal stability. Bias-cut edges eliminate potential for raveling.
Suitable for ISO Class 6 (FED-STD-209E Class 1000/M4.5) environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty