Would you like to log in again?
Pro-Wipe® 880 Polypropylene Multi-Layer Nonwoven Cleanroom Wipes, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PW8801212.12 PW8800810.12
21914-356CS 640.5 USD InStock
21914-356 21914-358
Pro-Wipe® 880 Polypropylene Multi-Layer Nonwoven Cleanroom Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Pro-Wipe® 880 is a thermally bonded multi-layer nonwoven wiper recommended for ISO class 5 and above environments.
  • Engineered for compatibility with acid solvents
  • Extremely durable and pure with very low particle and fiber generation
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
Engineered for compatibility with acid solvents. Chemically compatible with common cleaning and disinfecting solutions. Excellent for general wiping and cleaning in biomedical environments.
This combination of properties provides with cleanliness and chemical resistance to strong solvents including acids.
Certifications: Certified for contact with food per EC Regulation 1935(2004) - FDA 21 CFR - Food contact compliant.
Ordering information: 100 sheets per pack, 12 packs per case (double packed).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty