Would you like to log in again?
Precision Clean® II Class 100 Cleanroom Bags, PPC Flexible Packaging
Supplier: PPC Flexible Packaging
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Precision Clean®
10425 STCK0002-001 STCK0014-001 STCK0004-001 VWRI0015-001 STCK0003-001 10299 10424 10060A 10082A STCK0007-001 10207 STCK0012-001 10287 STCK0020-001 STCK0069-001 10580A STCK0011-001 STCK0025-001 10048 10049A STCK0010-001 VWRI0010-001 STCK0005-001 10171
21915-280CS 816.39 USD InStock
21915-280 21915-284 21915-042 21915-140 21915-040 21915-060 21915-115 21915-316 21915-318 21915-058 21915-056 21915-310 21915-312 21915-170 21915-304 21915-306 21915-308 21915-046 21915-100 21915-166 21915-122 21915-066 21915-044 21915-164 21915-302
Precision Clean® II Class 100 Cleanroom Bags, PPC Flexible Packaging
Bags Sample Bags
These bags are ideal for packaging applications requiring ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions.
They are made of clear polyethylene. Choose from 2, 4, or 6 mil thickness.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty