Would you like to log in again?
Hazmax® Knee Boots, Onguard
Supplier: Onguard
Hazmax®
87050-12 87050-09 87050-14 87050-10 87050-13 87050-11
21921-796PK 188.46 USD InStock
21921-796 21921-798 21921-788 21921-792 21921-794 21921-790
Hazmax® Knee Boots, Onguard
Shoes and Boots
One-piece, seamless, injection-molded PVC/Urethane over shoe 43.2 cm (17"). Fits over most work shoes and boots.
  • Nonconductive adjustable polyester strap with non-metallic buckle and snaps
  • High-visibility green upper with black, easy-to-clean, nonslip cleated sole
  • Heel kick-off lug for easy removal
  • Available in men’s sizes only
Boot provides maximum resistance to chemicals and hazardous materials.
Color: Green
Caution: Warning: Not for use for fire protection.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty