Would you like to log in again?
Slicker Boots, Overboots with Strap, Onguard
Supplier: Onguard
88050-10 88050-13 88050-11 88050-14 88050-12
21921-812CS 653.04 USD InStock
21921-812 21921-810 21921-808 21921-804 21921-806
Slicker Boots, Overboots with Strap, Onguard
Shoes and Boots
Injected molded seamless construction made from PVC / FLEX-O-THANE compound. Fit over most work shoes and boots.
  • Excellent cold weather flexibility
  • Non-absorbent polyester liner for easy cleaning and quick drying
  • Non-conductive, adjustable polyester strap with plastic buckle and snap for better fit
  • Self-cleaning, cleated sole
  • Kick-off lug for easy boot removal
Complete waterproof protection plus high chemical resistance. Excellent for use in construction, concrete work, utilities and food processing
Color: Yellow
Height: 41 cm (16")
Ordering information: When ordering, order by shoe size.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty