Would you like to log in again?
Hazmat Boot Covers, Onguard
Supplier: Onguard
97591-L 97591-XL
21921-844CS 642.6 USD InStock
21921-844 21921-846
Hazmat Boot Covers, Onguard
Disposable Garments
These latex boot covers offer protection from a broad range of materials, both hazardous and non hazardous.
  • High visibility yellow color
  • Strong enough to be reusable, economical enough to be disposable
  • Ribbed and textured sole provides safe, dependable traction
Made of 0.7 mm latex rubber, these seamless, waterproof covers are designed to provide a snug fit over existing footwear for an added layer of chemical and wear protection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty