Would you like to log in again?
Hazmax® Hazardous Material Boots, Onguard
Supplier: Onguard
Hazmax®
87012-11 87012-07 87012-08 87012-12 87012-13 87012-10 87012-15 87012-14 87012-06 87012-09
21921-976CS 194.39 USD InStock
21921-976 21921-965 21921-978 21921-968 21921-972 21921-962 21921-974 21921-964 21921-970 21921-960
Hazmax® Hazardous Material Boots, Onguard
Shoes and Boots
The boot that continues to set the standard for biological hazards and waste protection.
  • Steel toe, steel shank, steel midsole with Ultragrip® sipe outsole
  • 16" boot height
  • Exceeds CSA standards for dielectric footwear
  • One-piece injection molded construction
  • Replaceable cushioned insoles
  • Cut off bands
  • Kick off lug
Hazmax® hazardous material boots are the finest boots made for hazardous material handling. They are manufactured from a high viscosity and flexible PVC alloy, and feature a non-absorbent polyester lining for easy cleaning and quick drying. The one-piece injection molded construction eliminates seams, while the steel toe, steel shank, and steel midsole offer complete protection. They will provide limited flash-fire protection (escape only) in the event of a chemical fire. Ideally suited for use in applications involving dangerous chemicals, gasses, or biological hazards and waste.
Certifications: Steel toe complies with ASTM F2413-11 and CSA Z195.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty