Would you like to log in again?
CHEMTEX COVERALL
Supplier: ONGUARD INDUSTRIES
Coverall^ Chemtex coverall with attached hood. Level C garment not to be used in Level A environments. Heavy duty PVC on high count nylon polyester (.35mm thickness). Reinforced knees, elbows and crotch area to prevent splitting. Small.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHEMTEX COVERALL 21922-146 71022-SM Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty