Would you like to log in again?
CHEMTEX BIB OVERALL
Supplier: ONGUARD INDUSTRIES
Bib Overalls^ Chemtex bib overall, plain front. Level C garment not to be used in Level A environments. Heavy duty PVC on high count nylon polyester (.35mm thickness). Reinforced knees, elbows and crotch areas to prevent splitting. Heavy duty adjustable suspender with deluxe suspender clip. Small.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHEMTEX BIB OVERALL 21922-174 71050-SM Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty