Would you like to log in again?
CHEMTEX BIB OVERALL
Supplier: ONGUARD INDUSTRIES
Bib Overalls^ Chemtex bib overall, plain front. Level C garment not to be used in Level A environments. Heavy duty PVC on high count nylon polyester (.35mm thickness). Reinforced knees, elbows and crotch areas to prevent splitting. Heavy duty adjustable suspender with deluxe suspender clip. 3X-Lrg.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHEMTEX BIB OVERALL 21922-184 71050-3X Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty