Would you like to log in again?
Sanitex Hood, Onguard
Supplier: Onguard
71260-LG
21922-238EA 14.53 USD InStock
21922-238
Sanitex Hood, Onguard
Workwear
Protection for all sanitation situations and needs.
  • Heavy-duty PVC on high count nylon polyester
  • Green finish
  • 0.35 mm material thickness
  • Cord locks on hood strings
The sanitex hood features 0.35 mm high density PVC on nylon polyester heavy-duty construction for an elongated life span. The garment features cord locks on hood strings and offers protection for all sanitation requirements. Ideally suited for use in field refinery, environmental clean up, chemical plant, and food processing applications.
Certifications: Complies with ASTM D6413 for flame resistance.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty