Would you like to log in again?
PORTA-VAC II WAND 4INWAFR TIP
Supplier: VIRTUAL INDUSTRIES
Porta-Vac* II Portable Vacuum Wand^ Cleanroom and ESD-safe. Molded peek tip handles up to 10.2cm wafers. Features a high-performance pump and standard rear exhaust. Battery-operated with no hoses or cords. Compact and lightweight. Flashing LED indicates low battery.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PORTA-VAC II WAND 4INWAFR TIP 21925-150 PV4000A-MW4 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty