Would you like to log in again?
Porta-Vac® II Portable Vacuum Wand Kits, Virtual Industries
Supplier: Virtual Industries
Porta-Vac®
PV4000A-MW6 PV4000A-MW8
21925-152EA 1854.05 USD InStock
21925-152 21925-154
Porta-Vac® II Portable Vacuum Wand Kits, Virtual Industries
Vacuum Handling Systems
Cleanroom and ESD-safe, the portable wafer wand features a high-performance pump and standard rear exhaust.
  • 9 volt alkaline battery operation
  • Class I cleanroom approved
  • ESD safe
  • No hoses to get in the way
  • Portable
  • Handles wafers, flat panels
Ideal for handling wafers, flat panels or any item that has a flat, hard surface. Easily lifts over 500 grams. Rear exhaust of the pump air is standard on these units. The battery low indicator flashes red when the battery needs to be changed. A large on/off switch provides easy on/off control. An internal air filter is replaceable from the front of the tool. The Class 1 exhaust filter ensures clean operation. This tool will accept any of our handling tips that are have the press-fit-adaptor.
Ordering information: For available accessories or replacement items, contact your VWR representative.
Delivery information: This handling tool kit comes with one Molded Peek™ wafer tip. A disposable 9V battery is included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty