Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Cup Kit, Virtual Industries
Supplier: Virtual Industries
V9000K8-B
21925-166EA 25.32 USD InStock
21925-166
Vacuum Cup Kit, Virtual Industries
Vacuum Handling Systems
High-quality ESD-safe vacuum cups for use in electronics/SMT, optics, mechanical assembly, semiconductor handling and robotics
Made of black Buna-n static dissipative material.
Ordering information: Supplied with one each of the following seven diameters: 3/32", 1/8", 1/4", 3/8", 1/2", 5/8", 3/4".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty