Would you like to log in again?
SPECTRO CELLS W/FUSDTOP 4MC PK1
Supplier: SCIENTIFIC CELLS
Spectrophotmeter Cell^ With fused top. Path length: 10mm. Volume: 4mL. For use with spectrophotometers.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SPECTRO CELLS W/FUSDTOP 4MC PK1 22200-350 110QS10MM4 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty