Would you like to log in again?
Super Erecta® Polymer Stem Casters, Metro™
Supplier: Metro International
Super Erecta™ Super Erecta®
5PC 5PC 5PCR 5PCB 5PCB 5PCR
22233-714EA 178.06 USD InStock
CA22233-712 22233-714 CA22221-206 22233-712 22221-206 CA22233-714
Super Erecta® Polymer Stem Casters, Metro™
Furniture Shelves
Use these stem casters with Super Erecta® shelving and SiteSelect® UP posts (see 22233-716 series) to create mobile units.
  • Innovative polymer stem casters offer corrosion-resistance and enhanced durability
  • Appropriate for all medium-duty mobile applications
Casters feature all-polymer horn, stainless steel axle and hardware, sleeve-style axle bearing and an optional toe-operated brake mechanism. They measure 12.7 cm (5") and are load rated at 135 kg (300 lbs.). Donut bumpers are included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty