Would you like to log in again?
Olympic™ DIP Tube Tote Boxes, Metro International
Supplier: Metro International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Olympic™
TB95050NAT CI95050CAS TB95050CAS TB95050NAT TB95050CAS CI95050CAS
22221-316EA 45.98 USD InStock
CA22221-316 CA82002-506 22221-316 CA22221-282 82002-506 22221-282
Olympic™ DIP Tube Tote Boxes, Metro International
Boxes ESD-Safe Boxes
These boxes offer safe and efficient storage of DIP tubes and other items
Natural boxes are made of a polypropylene-based resin material and have a high chemical resistivity. Natural boxes are gray. Bentron™ is a polypropylene-based material with a carbon additive to provide an electrically conductive path through the material. Bentron™ has a surface resistivity of <105ohms/sq. Bentron™ products are black.
The insert style cover fits inside the rim of the tote box and has a finger hole that enables easy removal.
Boxes are durable, stackable, and feature high impact strength. There are flat surfaces on each side of the box that are convenient places to affix labels and bar codes. Choose from Natural or Bentron™ material.
Ordering information: Dividers, card holders, other sizes of tote boxes, and boxes and covers made of other materials are available. Contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty