Would you like to log in again?
Bentron™ Conductive Tote Boxes, Metro International
Supplier: Metro International
Olympic™ Bentron™
TB91035CAS TB93030CAS TB92050CAS TB93060CAS TB93080CAS TB92060CAS TB93080CAS TB93120CAS TB92080CAS TB91060CAS TB91060CAS TB92050CAS TB92060CAS TB91035CAS TB92035CAS TB93030CAS TB93060CAS TB93120CAS TB92035CAS TB92080CAS TB93050CAS TB93050CAS
22221-292EA 93.48 USD InStock
22221-292 CA22221-290 22221-290 CA22221-246 CA22221-244 CA22221-288 22221-246 22221-284 22221-296 CA22221-294 CA22221-292 22221-294 CA22221-286 22221-244 CA22221-242 22221-288 CA22221-298 22221-286 22221-242 CA22221-284 CA22221-296 22221-298
Bentron™ Conductive Tote Boxes, Metro International
Boxes ESD-Safe Boxes
Tote boxes offer safe and efficient storage of items in bulk or in compartmentalized fashion with the use of optional dividers and covers
These conductive tote boxes are made of polypropylene.
Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes. ESD Classification: Conductive.
Ordering information: Totes are also available in natural gray and blue static dissipative Benstat™. Contact your Avantor representative for more information.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty