Would you like to log in again?
SHELF GRID WORKBENCH
Supplier: Metro
Grid Shelf^ (Pba-Gsd) For Use With Metro Smartbench* Esd Workbenches (22221-504, -506, -508, -338). Mounts To The Wire Grid Panel (22221-376, -378). Black Powder Coating. Dimensions: 22.9Wx47Lcm. Load Capacity: 22.7Kg. Shipping Weight: 1.8Kg.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHELF GRID WORKBENCH 22221-388 PBA-GSD Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty