Would you like to log in again?
Super Erecta Cart Covers, Conductive, Metro™
Supplier: Intermetro
Super Erecta®
18X36X62ESD 24X48X62ESD 24X36X62ESD 18X48X62ESD 18X36X62ESD 18X48X62ESD 24X36X62ESD 24X48X62ESD
22221-446EA 909.23 USD InStock
CA22221-444 22221-446 22221-444 CA22221-438 CA22221-446 22221-438 CA22221-440 22221-440
Super Erecta Cart Covers, Conductive, Metro™
Covers
ESD Cart Covers provide an effective method of controlling electrostatic discharge while portecting cart contents from contamination and moisture
All ESD Covers inlcude a flap on one side, zipper closures and a paperwork pouch sized to accommodate 8 1/2 x 11" (2126 x 729 mm) documents.
ESD Classification: Conductive outer surface and static dissipative inner surface.
Covers feature a slip resistant texture which leaves no black carbon residue. Exterior color is black.
Certifications: All items with an ESD classification of conductive or static dissipative conform to the standards and testing procedures as prescribed per: EIA Standard 541, MIL-HDBK-263A and EOS/ESD-Sll.ll-1993.
Ordering information: Covers are designed to fit standard Metro™ carts with 63" Post Heights. Other sizes are available. Contact your Avantor representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty