Would you like to log in again?
Super Erecta Shelf® Ledges, Side and Back, 25 mm (1"), Metro™
Supplier: Metro International
Super Erecta® Super Erecta Shelf®
L54N-1C L48N-1S L14N-1C L18N-1C L42N-1S L72N-1C L36N-1C L30N-1C L24N-1C L60N-1C L72N-1S L60N-1C L30N-1S L30N-1S L72N-1C L48N-1C L42N-1C L18N-1S L60N-1S L54N-1C L36N-1S L18N-1C L42N-1S L21N-1C L30N-1C L42N-1C L18N-1S L14N-1S L48N-1C L36N-1S L72N-1S L48N-1S L21N-1S L24N-1S L24N-1C L21N-1C L54N-1S L54N-1S L14N-1S L24N-1S L60N-1S L14N-1C L36N-1C L21N-1S
22233-248EA 55.25 USD InStock
22233-248 CA22233-234 CA22233-256 22233-246 22233-268 CA22233-232 CA22233-254 CA22233-238 CA22233-258 CA22233-236 22233-240 22233-262 CA22233-270 CA19766-291 22233-260 22233-244 22233-266 CA22233-252 22233-242 22233-264 CA22233-250 CA19766-013 CA22233-246 CA22233-268 22233-236 22233-258 CA22233-244 CA22233-266 22233-234 22233-256 CA22233-248 22233-238 CA22233-260 22233-250 19766-291 22233-270 CA22233-264 CA22233-242 22233-254 22233-232 CA22233-262 CA22233-240 22233-252 19766-013
Super Erecta Shelf® Ledges, Side and Back, 25 mm (1"), Metro™
Furniture Shelves
Super Erecta Shelf® Ledges for use with Super Erecta® shelving systems.
  • For stationary or mobile installations
  • Ledges prevent items from protruding or falling from shelves
Note: Not stackable. Actual ledge length is approximately 2.5 cm (1") shorter than the nominal shelf length/width.
Ordering information: For designer color ledges, color must be specified at time of order. Designer colors include black matte, flame, hunter green, copper hammertone, and silver hammertone. Contact your VWR representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty