Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Super Erecta Shelf® Ledges, 100 mm (4"), Stackable, Metro™

Supplier: Metro International
Super Erecta®
L54N-4C L30N-4C L60N-4C L14N-4S L30N-4C L54N-4S L24N-4C L72N-4S L30N-4S L21N-4C L30N-4S L48N-4S L54N-4S L21N-4S L42N-4C L42N-4S L18N-4S L24N-4S L48N-4C L42N-4S L36N-4S L54N-4C L60N-4S L21N-4S L72N-4C L18N-4C L36N-4C L48N-4S L42N-4C L60N-4S L24N-4C L24N-4S L18N-4C L36N-4C L14N-4C L14N-4S L36N-4S L72N-4S L60N-4C L72N-4C L14N-4C L21N-4C L18N-4S L48N-4C
22233-302EA 121.75 USD InStock
CA22233-312 22233-302 CA22233-278 CA22233-310 CA22233-298 22233-300 CA22233-276 22233-306 CA22233-314 22233-304 22233-284 CA22233-292 22233-282 CA22233-290 22233-288 CA22233-274 CA22233-296 22233-286 CA22233-294 CA22233-272 22233-280 CA22233-308 CA22233-306 22233-314 CA22233-300 CA22233-288 22233-312 22233-278 CA22233-304 CA22233-302 CA22233-282 22233-294 22233-272 CA22233-280 22233-292 22233-310 CA22233-286 22233-276 22233-298 CA22233-284 22233-296 22233-274 22233-290 22233-308
Super Erecta Shelf® Ledges, 100 mm (4"), Stackable, Metro™
Furniture Shelves
Stackable Super Erecta Shelf® Ledges for use with Super Erecta® shelving systems.

  • May be stacked to form an 8" (203 mm) or higher barrier
  • Ledges prevent items from protruding or falling from shelves

Note: Actual ledge length is approximately 2.5 cm (1") shorter than the nominal shelf length/width.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty