Would you like to log in again?
Axygen® MAXYMum Recovery™ Hydrophobic Filter Barrier Pipette Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen® Maxymum Recovery™
TF-400-L-R-S TF-100-L-R-S TF-420-L-R-S TF-200-L-R-S TXLF-10-LRS TF-20-L-R-S TF-50-L-R-S TF-300-L-R-S TF-1000-L-R-S TF-350-L-R-S
22234-024CS 1050.24 USD InStock
22234-024 22234-016 47749-868 22234-018 22234-008 22234-010 22234-020 22234-012 47749-870 22234-132
Axygen® MAXYMum Recovery™ Hydrophobic Filter Barrier Pipette Tips, Corning
Pipette Tips
For applications that require high accuracy and reproducibility.
  • Compatible with most major brands of pipettors
Manufactured using diamond-polished molds to eliminate the surface cavities and occlusions which cause sample retention. Polymer resists DNA, RNA, and protein adherence and will not denature DNA. Manufactured without secondary substances such as silicone or heavy metals, which can contaminate samples.
Tips are autoclavable and presterilized through exposure to e-beam radiation for protection from contamination.
Ideal for DNA multiplication and handling radioactive, biohazardous, and corrosive samples. Filter prevents cross-contamination from the pipette to subsequent samples.
Certifications: Certified to be free of DNase, RNase, and pyrogens.
Ordering information: The 10 to 300 µl tips and 1250 µl tips 12000-756 are packaged 96 per sealed-bottom hinged rack, 500 and 1000 µl tips are packaged 100 per non-hinged rack. For non sterile tips, contact your sales representative.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty