Would you like to log in again?
Axygen® MAXYMum Recovery™ Research-Grade Pipette Tips, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen® Maxymum Recovery™
TXL-10-L-R T-300-L-STK TR-222-C-L T-400-L-R T-1000-C-L T-200-C-L-STK TXL-10-L T-400-L T-400-L-R-S TR-222-C-L-R-S T-200-C-L-R T-1000-C-L-R-S T-200-C-L-R-S T-300-L TXL-10-L-R-S T-1000-C-L-R T-350-C-L T-350-C-L-R-S T-300-L-R T-350-C-L-R TR-222-C-L-STK T-400-R-S TR-222-C-L-R T-300-L-R-S T-200-C-L
22234-090CS 594.28 USD InStock
22234-090 22234-070 22234-092 12000-210 12000-212 22234-094 22234-072 22234-074 22234-096 22234-098 22234-076 22234-130 22234-102 22234-068 22234-128 10011-872 10011-011 22234-062 22234-086 12000-208 22234-064 22234-100 CA10011-347 22234-066 22234-088
Axygen® MAXYMum Recovery™ Research-Grade Pipette Tips, Corning
Pipette Tips
For applications requiring high accuracy and reproducibility.
  • Ideal for transferring viscous DNA, RNA, and protein solutions
  • Autoclavable under normal autoclaving guidelines
Tips are constructed using diamond-polished molds to eliminate the cavities and occlusions that cause sample retention. Made of a unique polymer that resists DNA, RNA, and protein adherence, tips will not denature DNA. Manufactured without any secondary substances such as fillers, silicone, or heavy metals, which can contaminate samples.
Presterilized tips are exposed to e-beam radiation for protection from contamination.
All tips feature the 'Uni-grip' design, which assures an airtight seal with most major brands of pipettors.
Certifications: Certified to be free of DNase, RNase, and pyrogens.
Ordering information: Graduated tips have graduations at 10, 50, and 100 µl. The 200 µl wide-bore tips have an orifice size of 1.52 mm (¹/₁₆"). Tips 12000-738, 12000-740, 47734-663, and 47734-665 are designed for use with 16-channel pipettors for either 384- or 1536-well pipetting.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty