Would you like to log in again?
PYREX® Cloning Cylinder, Corning
Supplier: Corning
PYREX®
3166-6 3166-8 3166-10
22877-254CS 104.03 USD InStock
22877-254 22877-256 22877-252
PYREX® Cloning Cylinder, Corning
Cylinders Cloning Cylinders
Cloning cylinders, made of borosilicate glass code 7740. Clone a single cell or group of cells by surrounding them with this open ended cloning cylinders. Dip the end of the cylinder into sterile silicone grease before pressing to the bottom of a culture dish to create an isolated well.
  • Reusable
  • Non sterile
  • Cylinders are autoclavable or can be dry heat sterilized
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty