Would you like to log in again?
Applicator Brushes, Gordon Brush
Supplier: Gordon Brush
1CKG-12 1HHG-12
22940-508EA 34.78 USD InStock
22940-508 22940-510
Applicator Brushes, Gordon Brush
Brushes
This applicator brush is an inexpensive utility duster/cleaner that is an excellent replacement for disposable acid brushes that quickly shed their fibers.
  • Adhesive-free, to prevent leaching and contamination of surface areas
  • Anti-static
Brush fibers are secured using a zinc plated steel handle, substituting adhesives with 5 tons of mechanical clamping.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty