Would you like to log in again?
Double-Ended Applicator Brushes, Gordon Brush
Supplier: Gordon Brush
1005HHG-12 1005CKG-12 1010HHG-12
22940-528EA 52.03 USD InStock
22940-528 22940-530 22940-532
Double-Ended Applicator Brushes, Gordon Brush
Brushes
Two-sided applicator brushes. On one side, the brushes have a short flat end, and on the other side they have an extra stiff chiseled end.
  • Anti-static
  • Fibers are mechanically clamped into the handle, eliminating adhesives and ensuring maximum fiber retention
  • Great alternative to hand-trimming acid brushes
Chiseled end reaches and scrubs tight corners. Short, flat end is ideal for heavy-duty scrubbing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty