Would you like to log in again?
SST Series Heat-Tolerant Brushes, Insulative, Gordon Brush
Supplier: Gordon Brush
SST6NG-12 SST2NG-12
22940-566EA 6.46 USD InStock
22940-566 22940-568
SST Series Heat-Tolerant Brushes, Insulative, Gordon Brush
Brushes
Nylon bristle applicator brushes with a lightweight stainless steel handle.
  • Nylon bristle material is FDA approved
  • Stainless steel handle is chemical resistant and non-magnetic
  • Maximum use temperature of 250°F for short term exposures and 200°F for continuous or long term exposures
Manufactured using strong mechanical clamping for maximum fiber retention, and is adhesive-free to prevent contamination on surface areas caused by leaching.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty