Would you like to log in again?
Heat-Tolerant/Cleanroom Applicator Brushes, Gordon Brush
Supplier: Gordon Brush
250NADG-12 375NADG-12 499NADG-12
22940-590EA 70.51 USD InStock
22940-590 22940-592 22940-588
Heat-Tolerant/Cleanroom Applicator Brushes, Gordon Brush
Brushes
Pencil shaped applicator brushes with rigid acetal handle and Nylon bristles.
  • Can withstand most sterilizing environments
  • Bristles are epoxy set with high temperature adhesive that is autoclavable
Brushes must be cleaned prior to cleanroom use.
The brush is typically used in situations where a round brush is needed to clean fine areas, and for applying liquids or fluids in a high temperature setting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty