Would you like to log in again?
Wood Scratch Brushes, Gordon Brush
Supplier: Gordon Brush
30BG-12 30B 30SSG-12 30SS 30CKG-12 36CKG 15B-003G-12 36HHG
22940-602EA 17.16 USD InStock
22940-602 76238-014 22940-538 22940-606 22940-540 76243-100 22940-546 22940-534
Wood Scratch Brushes, Gordon Brush
Brushes
Antistatic brushes are ideal for cleaning circuit boards and SMT assemblies.
  • Plywood handles
  • Different versions available
One row brush has extra-fine, brass-fill fibers hand laced for soft cleaning, maximum retention, and durability.
The three row brush is small and economical with “scratch” or “toothbrush” style bristles.
The larger sratch brush has four rows of bristles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty