Would you like to log in again?
Plater's Brush, Gordon Brush
Supplier: Gordon Brush
444B006G
22940-642EA 51.32 USD InStock
22940-642
Plater's Brush, Gordon Brush
Brushes
This Plater's Brush is made with 0.006" Carbon Steel fill material and a hardwood shoe handle, contoured for a comfortable grip.
  • Recommended for use on plated parts and for fine metal finishing
  • High-quality brush with a 3 x 16 configuration
  • 1-1/8" trim, brush face of 4-3/4" x 5/8", handle length 10" x 1" wide
Platers' Brushes are designed for hand brushing, plating and fine metal finishing. The fill materials are softer than those used in scratch brushes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty