Would you like to log in again?
Horsehair and Wood Handle Dope Brush, Gordon Brush®
Supplier: Gordon Brush
DB7
22940-732EA 1.46 USD InStock
22940-732
Horsehair and Wood Handle Dope Brush, Gordon Brush®
Brushes
Large applicator dope brush with black horsehair bristles, clamped into a metal ferrule, with a sanded wood handle.
  • Economic and commonly used for pipe dope
Also recommended for gluing, pasting, cleaning up after soldering, and as a large acid flux applicator brush
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty