Would you like to log in again?
Sash Brush, Gordon Brush
Supplier: Gordon Brush
900046
22940-748EA 49.79 USD InStock
22940-748
Sash Brush, Gordon Brush
Brushes
This Size 4 hog bristle and plastic handle sash brush has an oval shape, and is excellent for dusting and removing debris.
  • Metal ferrule
  • Large tapered plastic handle
This useful brush has a trim length of 1-7/8", an overall length of 9-1/2" x 3/4" and a brush face width of 1-1/8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty