Would you like to log in again?
Counter Duster Brushes, Gordon Brush
Supplier: Gordon Brush
CDT CDHH CDHD CDFP
22940-760EA 102.64 USD InStock
22940-760 22940-762 22940-764 22940-766
Counter Duster Brushes, Gordon Brush
Brushes
All fill materials on these duster brushes are staple set into a clear, lacquered hardwood handle with a hang-up hole
For fine dusting, choose the horsehair (22940-760) or flagged plastic (22940-762) bristle brush.
For medium dusting, use the black tampico brush (22940-764).
The heavy-duty mill duster (22940-766) with brown plastic bristles has no side flare and a heavily filled front for aggressive dusting.
Brushes feature 5 rows of bristles widthwise and 15 rows of bristles lengthwise.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty