Would you like to log in again?
Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Supplier: Environmental Express
BPC3240
23050-048CS 54.02 USD InStock
23050-048
Pre-Cleaned Straight-Sided Jars, Polyethylene, Environmental Express
Jars General Use Jars
Natural high-density poylethylene jars include a polypropylene cap and F-217 PE liner.
All containers meet one of the three levels of pre-cleaning. Level III indicates that containers have not received an EPA washing treatment. Level II indicates that each case of containers has been custody-sealed and lot number-labeled for traceability to the enclosed Statement of Compliance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty