Would you like to log in again?
Pre-Cleaned Glass Jars with Open-Top/Septa Caps, Clear and Amber, Environmental Express®
Supplier: Environmental Express
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
APC3994 APC1930 APC3930
23050-188CS 139.38 USD InStock
23050-188 23050-172 23050-205
Pre-Cleaned Glass Jars with Open-Top/Septa Caps, Clear and Amber, Environmental Express®
Jars General Use Jars
Septa top offers easy access to contents through aspiration.
  • All containers meet one of two levels of pre-cleaning.
Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody sealed, lot number labeled for traceability, and a Certificate of Analysis for EPA quality assurance treatment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty